missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate trihydrate, 99+%, ACS reagent

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol

Supplier:  Thermo Scientific Chemicals 423840025

 View more versions of this product

Catalog No. AC423840025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) acetate trihydrate
6080-56-4
Crystalline Powder and/or Crystals
99.0 to 103% (Complexometry)
(CH3CO2)2Pb·3H2O
01,532; 02,233; 04,276
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead(2+);diacetate;trihydrate
22456
379.33
ACS Reagent
Decomposes information: >100°C
75.0°C
White
2.5 kg
C4H12O7Pb
MFCD00150023
15, 5452
Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
379.30
CHEBI:33112
≥99%
Plastic bottle
13.1

Chemical Identifiers

6080-56-4
379.30
MCEUZMYFCCOOQO-UHFFFAOYSA-L
22456
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
C4H12O7Pb
MFCD00150023
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
CHEBI:33112
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child.
Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHA

GHS Signal Word: Danger

RTECSNumber : OF8050000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only