missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate basic, ACS reagent

CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol

$136.44 - $136.44

Chemical Identifiers

CAS 51404-69-4
Molecular Formula C4H10O8Pb3
Molecular Weight (g/mol) 807.69
InChI Key XAOPZCQFBUCRKR-UHFFFAOYSA-L
PubChem CID 75087083
IUPAC Name diacetyloxylead;lead;dihydrate
SMILES CC(=O)O[Pb]OC(=O)C.O.O.[Pb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC453220010
SDS View Documents
Thermo Scientific Chemicals
453220010
1 kg
Each for $136.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51404-69-4
807.69
75087083
CC(=O)O[Pb]OC(=O)C.O.O.[Pb]
C4H10O8Pb3
XAOPZCQFBUCRKR-UHFFFAOYSA-L
diacetyloxylead;lead;dihydrate

Specifications

51404-69-4
C4H10O8Pb3
CC(=O)O[Pb]OC(=O)C.O.O.[Pb]
807.69
807.69
ACS Reagent
Lead(II) acetate basic
1 kg
XAOPZCQFBUCRKR-UHFFFAOYSA-L
diacetyloxylead;lead;dihydrate
75087083
≥33.0% (basic Pb as PbO)
1.5% max. (105°C, 2 h)
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Danger

EINECSNumber : 257-175-3

SDS
Documents

Documents

RUO – Research Use Only