accessibility menu, dialog, popup

UserName

H3COOPbOOCH3H2OH2OPb

CAS RN 51404-69-4

IUPAC Name: (acetyloxy)plumbyl acetate dihydrate lead
Synonyms: lead acetate (II), trihydrate dihydrate lead
Molecular Weight (g/mol): 568.5
Molecular Formula: C4H10O6Pb2
InChi Key: XAOPZCQFBUCRKR-UHFFFAOYSA-L
SMILES: O.O.[Pb].CC(=O)O[Pb]OC(C)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)