missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate basic, ACS reagent

Catalog No. AC453220050
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453220050 5 kg
AC453220010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453220050 Supplier Thermo Scientific Chemicals Supplier No. 453220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51404-69-4
Molecular Formula C4H10O8Pb3
Molecular Weight (g/mol) 807.69
InChI Key XAOPZCQFBUCRKR-UHFFFAOYSA-L
PubChem CID 75087083
IUPAC Name diacetyloxylead;lead;dihydrate
SMILES CC(=O)O[Pb]OC(=O)C.O.O.[Pb]

Specifications

Quantity 5 kg
Formula Weight 807.69
Percent Purity ≥33.0% (basic Pb as PbO)
Grade ACS Reagent
Loss on Drying 1.5% max. (105°C, 2 h)
Chemical Name or Material Lead(II) acetate basic
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed. Harmful if inhaled. May damage the unborn child. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray.
EINECSNumber 257-175-3

RUO – Research Use Only