missing translation for 'onlineSavingsMsg'
Learn More

Lead ICP Standard, 1000 ppm Pb in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PPB1KN100

 View more versions of this product

Catalog No. PPB1KN-100

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lead ICP Standard
7439-92-1,7697-37-2,7439-92-1,7732-18-5
93.0,4.89,0.09
1 mL = 1 mg Pb
Pb
Colorless
Approximately 0°C
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
100 mL
CHEBI:27889
Natural LDPE Bottle
1,000ppm Pb
97.0,5.09,0.1
Liquid
MFCD00134050
< 2
Standard
[Pb]
207.20
5352425

Chemical Identifiers

7439-92-1
207.20
WABPQHHGFIMREM-UHFFFAOYSA-N
CHEBI:27889
[Pb]
Pb
MFCD00134050
5352425
lead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions