missing translation for 'onlineSavingsMsg'
Learn More

Lead Free Paint - Powdered, MilliporeSigma™ Supelco™

Catalog No. 111001774
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001774 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001774 Supplier Merck Emd Millipore Supplier No. CRM05020G
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

pH 7.50
Type Certified Reference Material
Quantity 20 g
Grade Certified Reference Material