Learn More
Micronova™ Lead-Free Cleanroom Autoclave Tape
Lead-free autoclave indicator tape is a proven method of positively confirming completion of the sterilization cycle in both gravity discharge and vacuum autoclaves.
$505.38 - $505.38
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance RoHS Compliant; ISO 9001 Certified Facility; Certificate of Conformance
Description Green indicator lines change to black when proper levels of steam pressure & temperature are achieved
Adhesive Type Synthetic
Coating Rubber, Synthetic Resins
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity  
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity  
19-116-745
View Documents
Micronova™
AUTOPC34
60 yds. x 0.75 in.
Case of 16 Each for $505.38
Only null left
Description
Description
Micronova Lead-Free Autoclave Indicator Tape is an easy to handle, saturated crepe paper tape with synthetic polyisoprene adhesive and green indicator lines that change to black when proper levels of steam pressure and temperature have been achieved.
  • Lead-free ink indicator lines cannot be rubbed off
  • Adheres on contact and stays in place through live steam pressure
  • For use in gravity discharge and vacuum autoclaves
  • Printed lines cannot be rubbed off and will not contaminate instruments
  • Removes quickly and easily
  • Easy to handle, Latex-free
  • Wound on a 3 in. plastic core for cleanroom use
  • Certifications: RoHS Compliant
Specifications
Specifications
Not Autoclavable
Green indicator lines change to black when proper levels of steam pressure & temperature are achieved
Rubber, Synthetic Resins
Roll
Steam Autoclave Tape
Indicator tape for steam sterilization cycles.
3 in. Plastic Core
20 lbs./in.
0.15 mm
55 m
RoHS Compliant; ISO 9001 Certified Facility; Certificate of Conformance
Synthetic
Matte
Saturated Crepe Paper
Sterile
P-52, P-52W Tape Dispensers
Yes
6.5 mil
60 yd.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'