missing translation for 'onlineSavingsMsg'
Learn More

Lead Carbonate, Powder, Reagent, ACS, Spectrum™ Chemical

Catalog No. p-7157658
Change view
Click to view available options
Quantity:
12 kg
125 g
2.5 kg
50 kg
500 g
6 x 500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18603761 125 g Poly Bottle
18603762 500 g Poly Bottle
18603764 2.5 kg Poly Bottle
18603763 6 x 500 g Poly Bottle
18603765 12 kg Poly Pail
18603766 50 kg Fiber Drum
6 options

Catalog No. 18603761

Supplier: Spectrum Chemical Mfg Cor L1045125GM

Only null left

L1045, 598-63-0, PbCO3

Spectrum™ Chemical Lead Carbonate, Powder, Reagent, ACS is a chemical compound that is created industrially from lead acetate and carbon dioxide. It is an important pigment used in paints, putty, and ceramics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
IUPAC Name λ²-lead(2+) carbonate
SMILES [Pb++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Grade ACS
Packaging Poly Bottle