missing translation for 'onlineSavingsMsg'
Learn More

Lead Carbonate, Powder, CP, Approx. 95%, Spectrum™ Chemical

Catalog No. p-7165772
Change view
Click to view available options
Quantity:
12 kg
2.5 kg
50 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18611215 500 g Poly Bottle
18611214 2.5 kg Poly Bottle
18611213 12 kg Poly Pail
18611216 50 kg Fiber Drum
4 options

Catalog No. 18611215

Supplier: Spectrum Chemical Mfg Cor L1049500GM

Only null left

Chemical Identifiers

CAS 598-63-0

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity ∼95%
Grade Chemically Pure
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.