missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate Cotton, Ricca Chemical

Catalog No. R4293900100
Change view
Click to view available options
Quantity:
100 g
Packaging:
White Poly Wide Mouth Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R4293900100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4293900100 Supplier Ricca Chemical Company Supplier No. R4293900100B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Arsenic Testing

Chemical Identifiers

CAS 6080-56-4 , 9004-34-6
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name λ2-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Crystals and/or Chunks
Quantity 100 g
Solubility Information Solubility in Water: Insoluble in water
Grade Laboratory
Odor Odorless
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Lead Acetate Cotton, For Arsenic Testing