missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Lead

$40.85 - $40.85

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S25383B
SDS View Documents
Fisher Science Education™
S25383B
Ea. Sheet - 6.5 x 6.5 x 0.03 in.
Each for $40.85
Case of 25 Each for $1,021.25
Only null left
 
Description

Description

This product is for educational use - no C of A is available.

Specifications

Chemical Identifiers

7439-92-1
207.20
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
CHEBI:27889
[Pb]

Specifications

Blue-White to Gray
11.3g/mL
Sheet - 6.5 x 6.5 x 0.03 in.
Pb
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
5352425
207.2
7439-92-1
Ea.
100% w/v
Pb
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
[Pb]
207.20
CHEBI:27889
Lead
Videos
Safety and Handling

Safety and Handling

ShelfLife : 5 years

SDS
missing translation for 'documents'

missing translation for 'documents'