missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Lead

Catalog No. s25383b
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
Ea.
Packaging:
Sheet - 6.5 x 6.5 x 0.03 in.
Catalog No. Quantity Packaging
S25383B Ea. Sheet - 6.5 x 6.5 x 0.03 in.
1 options

Catalog No. S25383B

Supplier: Fisher Science Education™ S25383B

Only null left

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Blue-White to Gray
Density 11.3g/mL
Quantity Ea.
Packaging Sheet - 6.5 x 6.5 x 0.03 in.
Assay Percent Range 100% w/v
Linear Formula Pb
Formula Weight 207.2
Chemical Name or Material Lead
ShelfLife 5 years