missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qorpak™ LDPE Cubitainer with White PP SturdeeSeal™
For Use With: For storage and transport of liquid; Closure Material: Polypropylene; Color: Natural
$353.42 - $353.42
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Closure Material | Polypropylene |
| Color | Natural |
| For Use With (Application) | For storage and transport of liquid |
| Material | LDPE |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
02-991-765
|
Qorpak™
PLC03458 |
4 L |
Case of 8 for $353.42
|
|
|||||
Description
- Designed to collapse as product is dispensed
- Reversible well design permits opening to be raised or depressed for pouring and storage
- Can withstand rigors of hazardous chemical packaging
- 128 and 640 oz. (3.7 and 19L) containers include heavy-duty corrugated outer carton
Specifications
| Not Autoclavable | |
| Natural | |
| LDPE | |
| Square |
| Polypropylene | |
| For storage and transport of liquid | |
| Cubitainer |