missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDOC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552186 Supplier Invitrogen™ Supplier No. PA552186
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SVMAELTLLR TRARIPGALQ ITPPISSITS NGTRPMTTPP TSLPEPFSGD PGRLAGFLMQ MDRFMIFQAS RFPGEAERVA FLVSRLTGEA EKWAIPHMQP DSPLRNNYQG F Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

LDOC1L gene ontology (GO) annotation includes protein binding.

Specifications

Antigen LDOC1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTL6
Gene Accession No. Q6ICC9
Gene Alias 6-Mar; BC058638; dJ1033E15.2; gag-like protein; LDOC1L; leucine zipper down-regulated in cancer 1 like; leucine zipper protein down-regulated in cancer cells-like; leucine zipper, down-regulated in cancer 1 like; leucine zipper, down-regulated in cancer 1-like; mammalian retrotransposon-derived protein 6; MAR6; MART6; protein LDOC1L; Protein Sushi-15E3; Retrotransposon Gag-like protein 6; Rtl6; Sushi-15E3
Gene Symbols RTL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LDOC1L. Recombinant protein control fragment (Product #RP-91552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less