missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LC3A Recombinant Rabbit Monoclonal Antibody (23GB1960)
GREENER_CHOICE

Catalog No. MA553184
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553184 100 μL
MA553185 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553184 Supplier Invitrogen™ Supplier No. MA553184
Only null left

Rabbit Recombinant Monoclonal Antibody

LC3A Recombinant Monoclonal Antibody for Western Blot, ICC/IF

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen LC3A
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1960
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP1LC3A
Gene Accession No. Q6XVN8, Q91VR7, Q9H492
Gene Alias 1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3a; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; map1lc3a; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; similar to microtubule-associated protein 1 light chain 3 alpha; zgc:77094
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP1LC3A (1-34AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362245, 66734, 84557
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less