missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LC3A Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565969
Description
LC3A Monoclonal Antibody for IHC (P), Western Blot
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.Spécifications
| LC3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6XVN8, Q91VR7, Q9H492 | |
| MAP1LC3A | |
| Recombinant Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a) (Met1-Gly120). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C2 | |
| Unconjugated | |
| MAP1LC3A | |
| 1010001H21Rik; 4922501H04Rik; ATG8E; Autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3; LC3a; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; MAP1ALC3; MAP1BLC3; map1lc3a; microtubule associated protein 1 light chain 3 alpha; Microtubule-asso; microtubule-associated protein 1 light chain 3 alpha; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; similar to microtubule-associated protein 1 light chain 3 alpha; zgc:77094 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100322893, 362245, 66734, 84557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |