missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521642
Description
Recommended positive controls: A431, H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The LBP protein is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene.Chemical Identifiers
| LBP | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| LBP | |
| acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88 | |
| LBP | |
| Primary | |
| Antigen affinity chromatography |
| 0.46 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P18428 | |
| 3929 | |
| Recombinant protein encompassing a sequence within the center region of human LBP. The exact sequence is proprietary. | |
| Antibody |
Specifications
| LBP | |
| Polyclonal | |
| Unconjugated | |
| LBP | |
| acute phase protein; binds bacterial LPS; BPI fold containing family D, member 2; Bpifd2; hypothetical protein LOC512242; LBP; lipopolysaccharide binding protein; lipopolysaccharide-binding protein; lipopolysaccharide-binding protein LPSBP; LPS-binding protein; LPSBP; Ly88 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.46 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P18428 | |
| LBP | |
| Recombinant protein encompassing a sequence within the center region of human LBP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |