missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Layilin Monoclonal Antibody (K94011_7A4), Invitrogen™

Catalog No. MA556089
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556089 200 μL
MA555709 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556089 Supplier Invitrogen™ Supplier No. MA556089
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KRKREQPDPS TKKQHTIWPS PHQGNSPDLE VYNVIRKQSE ADLAETRPDL KNISFRVCSG EATPDDMSCD YDNMAVNPSE SGFVTLVSVE SGFVTNDIYE FSPDQMGRSK

Layilin is a cell surface receptor encoded by the LAYN gene, known for its involvement in cell adhesion and motility processes. It is a member of the C-type lectin-like domain protein family and plays a crucial role in mediating interactions with the extracellular matrix. Layilin is associated with cytoskeletal reorganization, impacting cellular processes such as shape change, migration, and stabilization of cell-matrix contacts. It has been shown to interact with other proteins like talin, contributing to the linkage between the cell membrane and the actin cytoskeleton, which is vital for maintaining cell structure and facilitating movement. Layilin's role in cell adhesion and motility makes it significant in developmental processes and wound healing. Additionally, dysregulation of layilin expression has been implicated in cancer metastasis, where altered adhesive properties and increased motility contribute to tumor progression and dissemination. Research is ongoing to explore the detailed mechanisms of layilin's function and its potential as a target for therapeutic intervention in diseases characterized by abnormal cell migration and adhesion.

Specifications

Antigen Layilin
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K94011_7A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene LAYN
Gene Accession No. Q6UX15
Gene Alias E030012M19Rik; FLJ30977; FLJ31092; Gm511; layilin; LAYN; LYAN; RGD1564444; UNQ208/PRO234
Gene Symbols LAYN
Host Species Mouse
Immunogen Recombinant human LAYN (immunogen range is 261-370).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less