missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lawesson's Reagent, 97%

Catalog No. p-7021973
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1453006 5 g
AAA1453014 25 g
AAA1453022 100 g
AAA1453036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1453006 Supplier Thermo Scientific Chemicals Supplier No. A1453006
Only null left

CAS: 19172-47-5 | C14H14O2P2S4 | 404.452 g/mol

Lawesson's reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lawesson′s reagent is a thiation agent used to convert carbonyl compounds into thiocarbonyls. It is also used to thionate enones, esters, lactones, amides, lactams and quinones. Further, it is used to prepare thiols from alcohols. It is associated with silver perchlorate and utilized as and oxophilic Lewis acid catalyst for Diels-Alder reaction of dienes with alfa, beta-unsaturated aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 19172-47-5
Molecular Formula C14H14O2P2S4
Molecular Weight (g/mol) 404.452
MDL Number MFCD00005171
InChI Key CFHGBZLNZZVTAY-UHFFFAOYSA-N
Synonym lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride
PubChem CID 87949
IUPAC Name 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
SMILES COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC

Specifications

Melting Point 221°C to 225°C
Quantity 5 g
UN Number UN3134
Beilstein 1024888
Sensitivity Moisture sensitive
Merck Index 14,5391
Solubility Information Insoluble in water.
Formula Weight 404.47
Percent Purity 97%
Chemical Name or Material Lawesson's Reagent
Hazard Category H261
Hazard Statement GHS H Statement
H331-H261-H302-H315-H319-H335
Toxic if inhaled.
In contact with water releases flammable gas.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: II
EINECSNumber 242-855-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only