Learn More
Lauryl chloride, 99%
CAS: 112-52-7 | C12H25Cl | 204.78 g/mol
Supplier: Thermo Scientific Chemicals 257100010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Lauryl chloride | |
| 112-52-7 | |
| Colorless | |
| 260°C | |
| Authentic | |
| Glass bottle | |
| 1.4420 to 1.444 | |
| MFCD00013688 | |
| 0.87 | |
| Solubility in water: 0.0068g/L (20°C). Other solubilities: miscible with most common organic solvents,slightly miscible with alcohol,soluble in ethanol,benzene,ether,miscible with acetone | |
| CCCCCCCCCCCCCl | |
| 204.78 | |
| 204.79 | |
| Liquid |
| 99% | |
| -9.3°C | |
| 0.8700g/mL | |
| 130°C | |
| 98.5% min. (GC) | |
| C12H25Cl | |
| CH3(CH2)11Cl | |
| 1 L | |
| lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
| YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
| 1-chlorododecane | |
| 8192 | |
| 99% |
Chemical Identifiers
| 112-52-7 | |
| 204.78 | |
| YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
| 8192 | |
| CCCCCCCCCCCCCl |
| C12H25Cl | |
| MFCD00013688 | |
| lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
| 1-chlorododecane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 203-981-5
TSCA : TSCA
RUO – Research Use Only