missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl-L-carnitine, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002216
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002216 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002216 Supplier Merck Emd Millipore Supplier No. 9143210MG
Only null left

Specifications

CAS 25518-54-1
Linear Formula C19H37 NO4
Synonym (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium inner salt; L-Carnitine dodecanoyl ester; C12-Carnitine; Dodecanoyl-L-carnitine
Type Analytical Standard
Quantity 10 mg
Formula Weight 343.50
Percent Purity ≥97% (HPLC)
Grade Analytical Standard