missing translation for 'onlineSavingsMsg'
Learn More

Lauric Anhydride 98.0+%, TCI America™

Catalog No. L001725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001725G 25 g
L0017100G 100 g
L0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L001725G Supplier TCI America Supplier No. L001725G
Only null left

Chemical Identifiers

CAS 645-66-9
Molecular Formula C24H46O3
Molecular Weight (g/mol) 382.629
MDL Number MFCD00008965
InChI Key NWADXBLMWHFGGU-UHFFFAOYSA-N
Synonym lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
PubChem CID 69527
IUPAC Name dodecanoyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 42°C
Color White
Quantity 25 g
Formula Weight 382.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lauric Anhydride