accessibility menu, dialog, popup

UserName

CAS RN 645-66-9

H3COOOCH3

CAS RN 645-66-9

IUPAC Name: dodecanoyl dodecanoate
Synonyms: dodecanoic acid, anhydride dodecenoyl anhydride
Molecular Weight (g/mol): 382.63
Molecular Formula: C24H46O3
InChi Key: NWADXBLMWHFGGU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)