missing translation for 'onlineSavingsMsg'
Learn More

Lauric acid, sodium salt, 98%

CAS: 629-25-4 | C12H23NaO2 | 222.30 g/mol

Supplier:  Thermo Scientific Chemicals 413040250

 View more versions of this product

Catalog No. AC413040250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lauric acid, sodium salt
629-25-4
Authentic
Glass bottle
CH3(CH2)10COONa
25 g
BTURAGWYSMTVOW-UHFFFAOYSA-M
sodium;dodecanoate
2735067
98%
99%
White to Yellow
98%
C12H23NaO2
MFCD00041754
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
[Na+].CCCCCCCCCCCC([O-])=O
222.30
222.3
Powder or Flakes

Chemical Identifiers

629-25-4
222.30
BTURAGWYSMTVOW-UHFFFAOYSA-M
2735067
[Na+].CCCCCCCCCCCC([O-])=O
C12H23NaO2
MFCD00041754
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
sodium;dodecanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-082-4

RTECSNumber : OF0700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only