accessibility menu, dialog, popup

UserName

CAS RN 629-25-4

IUPAC Name: sodium dodecanoate
Synonyms: sodium laurate
Molecular Weight (g/mol): 222.3
Molecular Formula: C12H23NaO2
InChi Key: BTURAGWYSMTVOW-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCC([O-])=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)