missing translation for 'onlineSavingsMsg'
Learn More

Lauric acid, sodium salt, 98%

Catalog No. AC413040050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413040050 5 g Glass bottle
AC413040250 25 g Glass bottle
AC413041000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC413040050 Supplier Thermo Scientific Chemicals Supplier No. 413040050

CAS: 629-25-4 | C12H23NaO2 | 222.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-25-4
Molecular Formula C12H23NaO2
Molecular Weight (g/mol) 222.30
MDL Number MFCD00041754
InChI Key BTURAGWYSMTVOW-UHFFFAOYSA-M
Synonym sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026
PubChem CID 2735067
IUPAC Name sodium;dodecanoate
SMILES [Na+].CCCCCCCCCCCC([O-])=O

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)10COONa
Quantity 5 g
Formula Weight 222.3
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Lauric acid, sodium salt, 99%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-082-4
RTECSNumber OF0700000
TSCA TSCA

RUO – Research Use Only