missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laser Printable Inserts

Catalog No. 19112769
Change view
Click to view available options
Color:
White
Size (Metric):
5 x 0.95 cm (W x H)
Size (English):
1.97 x 0.375 in. (W x H)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size (Metric) Size (English)
19112769 White 5 x 0.95 cm (W x H) 1.97 x 0.375 in. (W x H)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19112769 Supplier Brady™ Supplier No. LAT176124

For laser printers. Brady Laser Printable Inserts are made of B-124 non adhesive paper and also used as 110 terminal block marking for telecommunications.

Specifications

Color White
Finish Matte
Material Paper (B-124)
Product Type Laser Printable Label
For Use With (Application) Cable and wire marking, Panel labeling, Voice and data identification
For Use With (Equipment) Laser Printers
Shape Rectangle
Size (English) 1.97 x 0.375 in. (W x H)
Size (Metric) 5 x 0.95 cm (W x H)