missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Large Vessel Endothelial Supplement (LVES)

Catalog No. A1460801
Encompass
Change view
Click to view available options
Quantity:
11 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1460801 11 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1460801 Supplier Gibco™ Supplier No. A1460801

Large Vessel Endothelial Supplement (LVES)

Gibco™ Large Vessel Endothelial Supplement (LVES) is a 50X, complete, ready-to-use media supplement optimized for use with Gibco™ Medium 200 and Medium 200PRF for culture of primary human umbilical vein endothelial cells (HUVEC), human aortic endothelial cells (HAEC), and human pulmonary artery endothelial cells (HPAEC). This blended supplement has been designed to deliver optimal cell growth and to enable superior cell performance for angiogenesis applications, such as cell migration and tube formation. LVES contains fetal bovine serum, hydrocortisone, human epidermal growth factor, basic fibroblast growth factor, heparin, and ascorbic acid.

  • Perform tube formation, cell invasion, cell migration, and cell proliferation functional assays
  • Achieve superior growth rates of endothelial cells compared to low serum growth supplement (LSGS)
  • Expand primary large vessel endothelial cells without genomic alteration

    Verified Performance in Key Angiogenesis Applications
    Large Vessel Endothelial Supplement is designed to deliver optimal performance in tube formation, cell invasion, cell migration, and cell proliferation assays when used in conjunction with Medium 200, human umbilical vein endothelial cells, and Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix. These items and a detailed protocol are available in a convenient Angiogenesis Starter Kit or they can be purchased separately.

    Maximum Primary Cell Growth with One Blended Supplement
    Large Vessel Endothelial Supplement is a complete, ready-to-use blend of growth supplements optimized to deliver>4 x 104 HUVEC per cm2 when used with Medium 200 or Medium 200PRF. Cell phenotype is retained and verified positive for von Willebrand factor (vWf) and CD31, negative for a-actin, and positive for dil-Ac-LDL uptake via ICC.

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
  • Specifications

    Content And Storage 1 bottle. Store in freezer (-5 to -30°C).
    Cell Type Endothelial Cells (Umbilical Vein, Pulmonary Artery, Aortic, Large Vessel)
    Classification Animal Origin
    Concentration 50 X
    Form Liquid
    Product Type LVES Supplement
    Serum Level Standard Serum
    Sterility Sterile
    Without Additives No Glutamine
    Purity or Quality Grade Research Grade
    Quantity 11 mL
    Shipping Condition Dry Ice
    Source Human
    Species Human
    Tested For Growth Performance Assay
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.