Learn More
Invitrogen™ LanthaScreen™ Trousse d’analyse TR-FRET BACE1
Description
BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides found in extracellular amyloid plaques of Alzheimer's disease (AD). In some cases, early-onset familial AD can be attributed to a 'Swedish' mutation in the amyloid precursor protein (APP), which dramatically enhances the cleavage of this protein by BACE1. This and other genetic and pathological evidence has led to therapeutic approaches that focus on the inhibition of BACE1 and other APP-cleaving enzymes, such as gamma-secretase.
Invitrogen's LanthaScreen™ TR-FRET BACE1 assay provides sensitive high-throughput screening for potential inhibitors of beta-secretase. The kit uses a terbium (Tb)-labeled anti-biotin antibody and a fluorescein-labeled BACE1-biotin substrate in a homogeneous TR-FRET assay format (Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET BACE1 Assay Kit contains BACE1 protein, fluorescently labeled BACE1 substrate, Tb-labeled anti-biotin antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
Spécifications
Spécifications
| Contenu et stockage | À conserver au congélateur (de -5 °C à -30 °C). |
| Aperçu général | LanthaScreen™ TR-FRET BACE1 Assay Kit, 800 x 20μL, Liquid, β-Secretase Assay, Used in Protein Assays and Analysis, Peptide-Based Substrate Properties, Beta Secretase (BACE1) Target Enzyme, TR-FRET Technique, Beta Secretase Substrate |
| Essai | β-Essai de la sécrétase |
| Gamme de produit | LanthaScreen |
| Type de produit | LanthaScreen™ TR-FRET BACE1 Assay Kit |
| Propriétés de substrat | Substrat à base de peptides |
| Type de substrat | Substrat bêta-sécrétase |
| Suffisant pour | 800 x 20μL |
| À utiliser avec (application) | TR-FRET |
| Quantité | Essais 800 x 20 μL |
| Afficher plus de résultats |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.