Learn More
Thermo Scientific™ LanthaScreen™ Thiol Reactive Tb Chelate
Catalog No. p-7076023
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3580 10 μg
PV3578 1 mg
PV3579 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV3580 Supplier Thermo Scientific™ Supplier No. PV3580
Only null left
LanthaScreen™ Thiol Reactive Tb Chelate
  • As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Thiol Reactive Tb Chelate labeling reagent is available for assay development
  • The thiol-reactive maleimide group can be conjugated to virtually any peptide or protein that contains one or more accessible thiol moieties, such as the thiol group found on a cysteine residue
  • Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity
  • See the user guide for an application-based protocol for cysteine-containing peptides or proteins
Specifications
Quantity 10 μg
Product Type Thiol Reactive Tb Chelate
Content And Storage LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day.
Shipping Condition Dry Ice
Product Line LanthaScreen
For Research Use Only. Not for use in diagnostic procedures.