missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-PY20 Antibody Kit DFS Item

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

Supplier:  Thermo Scientific™ PV3552

 View more versions of this product

Catalog No. PV3552

Only null left
Explore available promotions

Description

Description

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Specifications

Tyrosine kinases
Tb (Terbium)/FITC/Lanthanide Chelates
25 μg
Kinase Antibodies and Immunoassays
1 Kit
150,000 Da
384-well Plate
Dry Ice
Signal Transduction
Tyrosine Kinases
ABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
0.45 mg/mL
150 kDa
RUO
Primary
Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C.
Biochemical terbium phospho-specific activity
340/490, 520 nm
End Point
ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, FGFR1, FGFR2, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, FLT1, MUSK, SRCN1, TEC, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, EGFR T790M, EPHA7, EPHB1, MET M1250T, FRK, ITK, FLT3 D835Y, SRC, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, ERBB2, NTRK2
TR-FRET
LanthaScreen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.