missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit

Catalog No. p-7181447
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3773 25 μg
PV3775 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3773 Supplier Invitrogen™ Supplier No. PV3773
Only null left

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents.

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.

In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.

Specifications

Description LanthaScreen™ Tb-labeled Anti-Rabbit Secondary Antibody Kit, 25μg, Liquid, Rabbit Species Reactivity, IgG Isotype, Terbium Conjugate, Fluorescent Detection Method, Water Soluble, 6 Months Shelf Life, Store in Freezer, Used for Pharma and Biopharm
Reactivity Rabbit
Detection Method Fluorescence
Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Dry Ice
Target Isotype IgG
Quantity 25 μg
Conjugate Lanthanide Chelates, Terbium (Tb)
Format Liquid
Formulation Liquid
Kit Contents LanthaScreen™ Tb-anti-Rabbit antibody, Fluorescein-Rabbit IgG, TR-FRET dilution buffer
For Use With (Application) Pharma and biopharma, secondary detection, target and lead identification and validation, protein biology, cell analysis, drug discovery and development, target and lead identification and validation, kinase biology, kinase antibodies and immunoassays, bi
Product Type LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.