missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-rabbit Ab

Catalog No. p-7181446
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3774 25 μg
PV3776 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3774 Supplier Invitrogen™ Supplier No. PV3774
Only null left

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies.

A general reagent for use in assay development, this terbium labeled antibody will bind rabbit antibodies. This product is ideally suited for use during assay development when evaluating multiple primary rabbit antibodies or when you are unable to directly terbium label your primary antibody.

Specifications

Description LanthaScreen™ Tb-labeled Anti-Rabbit Secondary Antibody, 25μg, Liquid, 1.03mg/mL Concentration, Rabbit Species Reactivity, Terbium Conjugate, Fluorescent Detection Method, 6 Months Shelf Life, Store in –20°C, Used for Pharma and Biophar
Reactivity Rabbit
Detection Method Fluorescence
Content And Storage Terbium-labeled anti-Rabbit antibody in HEPES buffered saline.

Store at -20°C.
Excitation/Emission 340/490, 520 nm
Readout End Point
Shipping Condition Dry Ice
Conjugate Lanthanide Chelates, Terbium (Tb)
Quantity 25 μg
Format Liquid
Formulation Liquid
For Use With (Application) Pharma and biopharma, secondary detection, cell analysis, drug discovery and development, target and lead identification and validation, kinase biology, kinase antibodies and immunoassays, biochemical kinase assays, secondary detection
Product Type LanthaScreen™ Tb-labeled Anti-rabbit Antibody
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.