missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit

Catalog No. p-7181445
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3765 25 μg
PV3767 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3765 Supplier Invitrogen™ Supplier No. PV3765
Only null left

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents.

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.

In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.

Specifications

Description LanthaScreen™ Tb-labeled Anti-Mouse Secondary Antibody Kit, 25μg, Liquid, Mouse Species Reactivity, Mouse Host, IgG Detection Target, Terbium Conjugate, Fluorescent Detection Method, Water Soluble, 6 Months Shelf Life, Store in Freezer, Used for
Content And Storage Store in freezer (-5°C to -30°C).
Reactivity Mouse
Shipping Condition Dry Ice
Quantity 25 μg
Detection Method Fluorescent
Conjugate Lanthanide Chelates, Terbium (Tb)
Target IgG
Format Liquid
Formulation Liquid
Kit Contents TR-FRET dilution buffer, LanthaScreen™ Tb-anti-mouse antibody, fluorescein-mouse IgG
For Use With (Application) Pharma and biopharma, secondary detection, target and lead identification and validation
Product Type LanthaScreen™ Tb-labeled Anti-Mouse Antibody Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.