missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-labeled Anti-Mouse Ab

Catalog No. p-7182869
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3766 25 μg
PV3768 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3766 Supplier Invitrogen™ Supplier No. PV3766
Only null left

LanthaScreen™ Tb-labeled Anti-Mouse Ab

A general reagent for use in assay development, this terbium labeled antibody will bind mouse antibodies. This product is ideally suited for use during assay development when evaluating multiple primary mouse antibodies or when you are unable to directly terbium label your primary antibody.

Specifications

Excitation/Emission 340/490, 520
Quantity 25 μg
Conjugate Lanthanide Chelates, Terbium (Tb)
Reactivity Mouse
Content And Storage Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C.
Readout End Point
Shipping Condition Dry Ice
Format Liquid

For Research Use Only. Not for use in diagnostic procedures.