missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Ab

Catalog No. p-7181443
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3772 1 mg
PV3770 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3772 Supplier Thermo Scientific™ Supplier No. PV3772

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is ideally suited for use during assay development when evaluating multiple primary goat antibodies, or when it is not possible to directly terbium label a primary antibody.

  • A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies
  • Ideally suited for use during assay development when evaluating multiple primary goat antibodies
  • Also for use when it is not possible to to directly terbium label a primary antibody

Specifications

Product Line LanthaScreen
Reactivity Goat
Content And Storage Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C.
Shipping Condition Dry Ice
Readout End Point
Excitation/Emission 340/490, 520
Quantity 1 mg
Conjugate Lanthanide Chelates, Terbium (Tb)

For Research Use Only. Not for use in diagnostic procedures.