Learn More
Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate
Description
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.
Specifications
Specifications
| Description | LanthaScreen™ Amine Reactive Tb Chelate, 10μg, 1050.88g/mol Molecular Weight, ≥90% by HPLC Purity, Store at –20°C, Used For Pharma and Biopharma, Target and Lead Identification and Validation |
| For Use With (Application) | Drug discovery & development, target & lead identification & validation, TR-FRET assay development |
| Quantity | 10 μg |
| Product Type | Amine Reactive Tb Chelate |
| Kit Contents | LanthaScreen™ TR-FRET labeling reagents are provided lyophilized |
| Molecular Weight (g/mol) | 1050.88g/mol |
| Purity | ≥90% by HPLC |
| Content And Storage | LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day. |
| Shipping Condition | Dry Ice |
| Product Line | LanthaScreen |
Frequently Asked Questions (FAQs)
We performed a comparison between the LanthaScreen assay and other commercially available TR-FRET assays from 2 different suppliers for the PKC kinase target. Our data revealed that the assays performed comparably, but that the LanthaScreen assay was simpler to optimize and contained fewer components that required optimization. The LanthaScreen assay is a two component system, whereas the other assay formats utilize a trimolecular mechanism which is more time consuming to optimize and has added costs.
Yes, this is possible depending on the concentrations of reagents used and the time for which they are mixed. We recommend developing and optimizing the assay by using separate reagent additions, because this method will work under the widest range of conditions. Once the assay is optimized, the performance of the assay using pre-mixed antibody and EDTA can be evaluated. We have successfully developed robust assays in which the antibody and EDTA were pre-mixed and then stored overnight at 4 degrees C prior to use the following day. A loss of signal intensity was observed in this case, however, by using the ratiometric readout, this effect was minimal.
The chelate is completely stable to Mg2+. The amount of Mn2+ or EDTA that the chelate can tolerate depends largely on how long they are mixed together and the combination of additives used in the reaction. If a reaction requires either Mg2+ or Mn2+ for activation, it is best to stop the reaction by adding an equimolar amount (or slight excess) of EDTA to chelate the metal ions present. This will then essentially eliminate any interference on the terbium chelate by EDTA or Mn2+. Regardless, when LanthaScreen assays are performed using a ratiometric readout (division of the acceptor signal by the donor signal), any interference caused by Mn2+ or EDTA is largely cancelled out.
The Förster radius, the distance at which energy transfer efficiency is half-maximal, is around 50-angstroms for the terbiumÆ fluorescein pair. However, the Förster radius does not give a complete indication of energy transfer efficiency when using long lifetime fluorophores such as terbium chelates. When using terbium chelates, energy transfer efficiency is determined by the distance of closest approach between the donor and acceptor during the excited state lifetime of the donor. In many assay systems, such as those designed using antibodies or peptides, there is a large degree of conformational freedom that allows the donor and acceptor to approach one another, effectively enhancing the FRET signal. Additionally, it is important to note that as the donor/acceptor pair approach one another and the efficiency of energy transfer increases, the fluorescent lifetime decreases to a comparable extent. From a practical standpoint, this means that when energy transfer is extremely efficient, FRET cannot be measured in time-resolved mode (because the energy transfer is complete before the measurement is made). This is another reason why TR-FRET assays based around terbium-labeled antibodies or streptavidin perform so well, because there exist a range of donor/acceptor distances, several of which are optimal for measuring FRET.
It varies, depending on the concentration of substrate used in the assay. But in general, for the peptide substrates, 1 mg of peptide will run approximately 250,000 wells (10 µL reaction, 200 nM peptide). For Poly GT or GAT, the 1 mL of 30 µM size we sell is approximately 1 mg. With these substrates, 1 mL of 30 µM will run approximately 16,700
wells (10 µL reaction, 200 µM substrate).
20 nmol of our physiological protein substrates is sufficient for approximately 10,000 wells (10 µL reaction, 200 µM substrate).
For Research Use Only. Not for use in diagnostic procedures.