missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfate hydrate, 99.9% (REO)

Catalog No. p-4824772
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4310622 100 g
AA4310636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4310622 Supplier Thermo Scientific Chemicals Supplier No. 04310622

CAS: 57804-25-8 | La2O12S3 | 565.98 g/mol

Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57804-25-8
Molecular Formula La2O12S3
Molecular Weight (g/mol) 565.98
MDL Number MFCD00011070
InChI Key VQEHIYWBGOJJDM-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) trisulfate
SMILES [La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point White heat (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Slightly soluble in water.
Formula Weight 566.00 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.821
Chemical Name or Material Lanthanum(III) sulfate hydrate
Show More Show Less
EINECSNumber 233-239-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only