accessibility menu, dialog, popup

UserName

SOOOOSOOOOSOOOOLaLa

CAS RN 57804-25-8

IUPAC Name: dilanthanum(3+) trisulfate
Synonyms: lanthanum sulfate
Molecular Weight (g/mol): 565.98
Molecular Formula: La2O12S3
InChi Key: VQEHIYWBGOJJDM-UHFFFAOYSA-H
SMILES: [La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results