missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) sulfate hydrate, 99.9% (REO)
Description
Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | White heat (decomposition) |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 99.9% (REO) |
| Density | 2.821 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 566.00 (anhydrous) |
| Show More |
Safety and Handling
| EINECSNumber | 233-239-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only