missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, 99%

CAS: 13709-38-1 | F3La | 195.90 g/mol

$89.32 - $278.92

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1667314
View Documents
Thermo Scientific Chemicals
A1667314
25 g
Each for $89.32
Only null left
 
AAA1667322
View Documents
Thermo Scientific Chemicals
A1667322
100 g
Each for $278.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13709-38-1
195.90
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
[F-].[F-].[F-].[La+3]
F3La
MFCD00011073
lanthanum(3+) trifluoride

Specifications

1493°C
25 g
LaF3
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
lanthanum(3+) trifluoride
195.9
Odorless
5.936
13709-38-1
F3La
MFCD00011073
[F-].[F-].[F-].[La+3]
195.90
99%
Hygroscopic
Lanthanum(III) fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-252-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only