missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, 99%

CAS: 13709-38-1 | F3La | 195.90 g/mol

Supplier:  Thermo Scientific Chemicals A1667322

 View more versions of this product

Catalog No. AAA1667322


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lanthanum(III) fluoride
13709-38-1
F3La
MFCD00011073
[F-].[F-].[F-].[La+3]
195.90
99%
Hygroscopic
1493°C
100 g
LaF3
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
lanthanum(3+) trifluoride
195.9
Odorless
5.936

Chemical Identifiers

13709-38-1
195.90
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
[F-].[F-].[F-].[La+3]
F3La
MFCD00011073
lanthanum(3+) trifluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-252-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only