missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Honeywell Fluka™

Catalog No. p-7122054
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
100 g
6 x 100 g
Packaging:
Ampoule
Poly bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6002390 100 g Poly bottle
60045978 6 x 100 g Poly bottle
6002403 1 Ea. Ampoule
60046051 6 x 1 Ea. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6002390 Supplier Honeywell Chemicals Supplier No. 31820100G
Only null left

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10025-84-0

Specifications

Quantity 100 g
Molecular Formula Cl3H14LaO7
Linear Formula LaCl3 · 7H2O
MDL Number MFCD00149756
UN Number NONH for all modes of transport
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) heptahydrate trichloride
Molecular Weight (g/mol) 371.36
Formula Weight 371.37g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Poly bottle
Chemical Name or Material Lanthanum(Iii) Chloride Heptahydrate
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
RTECSNumber OE4375000