Learn More
Lanthanum(III) chloride heptahydrate, 99%
CAS: 10025-84-0 | Cl3H14LaO7 | 371.36 g/mol
Supplier: Thermo Scientific Chemicals A155750E
Description
Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.
Solubility
Soluble in ethanol and water.
Notes
Incompatible with oxidizing agents, strong acids, strong reducing agents and active metals.
Specifications
| Lanthanum(III) chloride heptahydrate | |
| 10025-84-0 | |
| Cl3H14LaO7 | |
| 14,5363 | |
| FDFPDGIMPRFRJP-UHFFFAOYSA-K | |
| lanthanum(3+) heptahydrate trichloride | |
| 371.37 (245.27 Anhydrous) | |
| Odorless |
| 91°C (decomposition) | |
| 2500 g | |
| MFCD00149756 | |
| Soluble in ethanol and water. | |
| O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] | |
| 371.36 | |
| 99% |
Chemical Identifiers
| 10025-84-0 | |
| 371.36 | |
| FDFPDGIMPRFRJP-UHFFFAOYSA-K | |
| O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] |
| Cl3H14LaO7 | |
| MFCD00149756 | |
| lanthanum(3+) heptahydrate trichloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
H290-H317-H318
EINECSNumber : 233-237-5
RTECSNumber : OE4470000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only