missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. L02945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02945G 5 g
L02941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02945G Supplier TCI America Supplier No. L02945G
Only null left

Chemical Identifiers

CAS 168106-26-1
Molecular Formula C6F18LaN3O12S6
Molecular Weight (g/mol) 979.312
InChI Key XQHMUWQUZHVCJH-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Lanthanum(III) Salt, Lanthanum(III) Triflimide
PubChem CID 129827648
IUPAC Name bis(trifluoromethylsulfonyl)azanide;lanthanum(3+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[La+3]

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 979.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lanthanum(III) Bis(trifluoromethanesulfonyl)imide