missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, 99.9%

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

Supplier:  Thermo Scientific Chemicals 315232500

 View more versions of this product

Catalog No. AC315232500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1000.0°C
860.0°C
10099-58-8
250 g
Cl3La
MFCD00011068
Solubility in water: soluble
[Cl-].[Cl-].[Cl-].[La+3]
245.26
99.9%
Lanthanum chloride
White
Powder
99.9%
LaCl3
15,5414
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.26
Glass bottle

Chemical Identifiers

10099-58-8
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00011068
lanthanum(3+) trichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 233-237-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only