missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, 99.9%

Catalog No. AC315230100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315230100 10 g Glass bottle
AC315230500 50 g Glass bottle
AC315232500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315230100 Supplier Thermo Scientific Chemicals Supplier No. 315230100
Only null left

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Boiling Point 1000.0°C
Melting Point 860.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9%
Linear Formula LaCl3
Merck Index 15,5414
Solubility Information Solubility in water: soluble
Formula Weight 245.26
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Lanthanum chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 233-237-5

RUO – Research Use Only