missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole 98.0+%, TCI America™

Catalog No. p-7131882
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02331G 1 g
L02335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02331G Supplier TCI America Supplier No. L02331G
Only null left

Chemical Identifiers

CAS 103577-45-3
Molecular Formula C16H14F3N3O2S
Molecular Weight (g/mol) 369.362
MDL Number MFCD00866873
InChI Key MJIHNNLFOKEZEW-UHFFFAOYSA-N
Synonym lansoprazole, prevacid, bamalite, monolitum, lansoprazol, agopton, limpidex, ogastro, lanzor, opiren
PubChem CID 3883
ChEBI CHEBI:6375
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CS(=O)C2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 1 g
Formula Weight 369.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Lansoprazole
RTECSNumber DD9487500
TSCA No
Recommended Storage Refrigerator