missing translation for 'onlineSavingsMsg'
Learn More

Lansoprazole, 98+%

Catalog No. AAJ62008ME
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62008ME 500 mg
AAJ6200803 1 g
AAJ6200806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62008ME Supplier Thermo Scientific Chemicals Supplier No. J62008ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A proton pump inhibitor | CAS: 103577-45-3 | C16H14F3N3O2S | 369.362 g/mol

A proton pump inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103577-45-3
Molecular Formula C16H14F3N3O2S
Molecular Weight (g/mol) 369.362
MDL Number MFCD00866873
InChI Key MJIHNNLFOKEZEW-UHFFFAOYSA-N
Synonym lansoprazole, prevacid, bamalite, monolitum, lansoprazol, agopton, limpidex, ogastro, lanzor, opiren
PubChem CID 3883
ChEBI CHEBI:6375
IUPAC Name 2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES CC1=C(C=CN=C1CS(=O)C2=NC3=CC=CC=C3N2)OCC(F)(F)F

Specifications

Melting Point 173°C to 175°C
Color White
Quantity 500 mg
Merck Index 14,5362
Formula Weight 369.36
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Lansoprazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DD9487500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only