missing translation for 'onlineSavingsMsg'
Learn More

Chlorhydrate de landiolol 98,0+%, TCI America™

Numéro de catalogue. p-7131913
Modifier l'affichage
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
L029650MG 50 mg
L0296250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. L029650MG Fournisseur TCI America Code fournisseur L029650MG

Identifiants chimiques

CAS 144481-98-1
Molecular Formula C25H40ClN3O8
Molecular Weight (g/mol) 546.058
MDL Number MFCD00911043
InChI Key DLPGJHSONYLBKP-IKGOIYPNSA-N
PubChem CID 46856312
IUPAC Name [(4S)-2,2-diméthyl-1,3-dioxolan-4-yl méthyl 3-[4-[(2S)-2-hydroxy-3-[2-(morpholine-4-carbonylamino)éthylamino]propoxy]phényl]propanoate; Hydron; Chlorure
SMILES [H+].CC1(OCC(O1)COC(=O)CCC2=CC=C(C=C2)OCC(CNCCNC(=O)N3CCOCC3)O)C.[Cl-]

Spécifications

Melting Point 126°C
Color Blanc
Quantity 50 mg
Formula Weight 546.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material Landiolol Hydrochloride
RTECSNumber DA8346700
TSCA No